missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC83 Monoclonal Antibody (3E3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523184
This item is not returnable.
View return policy
Description
Peptide Sequence: SSDESTILHL SHENSIEDLQ YVKIDKEENS GTEFGDTDMK YLLYEDEKDF KDYVNLGPLG VKLMSVESKK MPIHFQEKEI PVKLYKDVRS PESHITYKMM KSFL.
CCDC83 (Coiled-Coil Domain Containing 83) is a Protein Coding gene.
Specifications
| CCDC83 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8IWF9 | |
| CCDC83 | |
| CCDC83 (NP_775827, 341 a.a. ~ 444 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3E3 | |
| Unconjugated | |
| CCDC83 | |
| 4930549K11Rik; 4930554C01Rik; 4932423M01Rik; CCDC83; coiled-coil domain containing 83; coiled-coil domain-containing protein 83; CT148; HSD9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 220047 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction