missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551695
This item is not returnable.
View return policy
Description
Immunogen sequence: MMSLLKDDVY CELAERHIQQ IVLFHQAGEE GGKVRRITSE GQILEQPLDP SLIPKLMSFL KLEKGKFGMV LLKKTLQVEE RYPYPVRLEA MYEVIDQGPI RRIEKIRQKG FVQKCKASGV EGQVVAEGND GGG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
CCDC80 (Coiled-Coil Domain Containing 80) is a Protein Coding gene. Diseases associated with CCDC80 include Herpes Zoster Oticus and Peripheral Nervous System Disease. Gene Ontology (GO) annotations related to this gene include heparin binding and glycosaminoglycan binding. Also, it is an estrogen regulated gene; overexpression correlates with mammary carcinogenesis.
Specifications
| CCDC80 | |
| Polyclonal | |
| Unconjugated | |
| CCDC80 | |
| 2610001E17Rik; Ccdc80; CL2; coiled-coil domain containing 80; coiled-coil domain-containing protein 80; coiled-coil domain-containing protein 80 pseudogene; down-regulated by oncogenes 1 protein; Down-regulated by oncogenes protein 1; DRO1; HBE245; LOW QUALITY PROTEIN: coiled-coil domain-containing protein 80; nuclear envelope protein okuribin; okuribin; Ssg1; SSG-1; steroid sensitive gene 1; steroid sensitive gene-1 protein; steroid-sensitive gene 1 protein; steroid-sensitive protein 1; up-regulated in BRS-3 deficient mouse; Up-regulated in BRS-3 deficient mouse homolog; Urb | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 151887 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q76M96 | |
| CCDC80 | |
| Recombinant protein corresponding to Human CCDC80. Recombinant protein control fragment (Product #RP-101003). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction