missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-50021
This item is not returnable.
View return policy
Description
CCDC8 Polyclonal specifically detects CCDC8 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| CCDC8 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| coiled-coil domain containing 8, coiled-coil domain-containing protein 8, DKFZp564K0322 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 83987 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q9H0W5 | |
| CCDC8 | |
| The immunogen for this product maps to a region between residue 350 and 400 of human Coiled-Coil Domain Containing 8 using the numbering given in entry NP_114429.2 (GeneID 83987). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction