missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCDC8 Polyclonal antibody specifically detects CCDC8 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CCDC8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | coiled-coil domain containing 8, coiled-coil domain-containing protein 8, DKFZp564K0322 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 488 and 538 of human Coiled-Coil Domain Containing 8 using the numbering given in entry NP_114429.2 (GeneID 83987). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?