missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC71 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28152-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| CCDC71 | |
| Polyclonal | |
| Unconjugated | |
| CCDC71 | |
| CCDC71, FLJ12800, FLJ13629 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 498678, 64925, 72454 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4V7C4, Q8IV32, Q8VEG0 | |
| Ccdc71 | |
| CCDC71 Fusion Protein Ag27446 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction