missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCDC69 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849082
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849082 Supplier Invitrogen™ Supplier No. PA520935

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

A suggested positive control is mouse lung tissue lysate. PA5-20935 can be used with blocking peptide PEP-1049.

May act as a scaffold to regulate the recruitment and assembly of spindle midzone components. Required for the localization of AURKB and PLK1 to the spindle midzone. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC69
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CCDC69
Gene Accession No. A6NI79, Q3TCJ8
Gene Alias 2210021E03Rik; CCDC69; coiled-coil domain containing 69; coiled-coil domain-containing protein 69; D11Ertd461e; RGD1562251
Gene Symbols CCDC69
Host Species Chicken
Immunogen A 16 amino acid peptide near the amino terminus of human CCDC69.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26112, 497906, 52570
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.