missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC68 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26301-1-AP-20UL
This item is not returnable.
View return policy
Description
Coiled-coil domain containing 68 (CCDC68) demonstrates a tumor-suppressive role in pancreatic ductal adenocarcinoma.Specifications
| CCDC68 | |
| Polyclonal | |
| Unconjugated | |
| CCDC68 | |
| CCDC68, CTCL associated antigen se57 1, SE57 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80323 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H2F9 | |
| CCDC68 | |
| CCDC68 Fusion Protein Ag23622 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction