missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC67 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552768
This item is not returnable.
View return policy
Description
Immunogen sequence: MKQNKVPRKE LPHLKEEIPF ELSNLNQKLE EFRAKSREWD KQEILYQTHL ISLDAQQKLL SEKCNQFQKQ AQSYQTQLNG KKQCLEDSSS EIPRLICDPD PNCEINERDE FIIEKLKSAV NEIALSRNKL QD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.
CCDC67 (deuterosome assembly protein 1) is a key structural component of the deuterosome, a structure that promotes de novo centriole amplification in multiciliated cells. Deuterosome-mediated centriole amplification occurs in terminally differentiated multiciliated cells and can generate more than 100 centrioles. Probably sufficient for the specification and formation of the deuterosome inner core. The protein interacts with CEP152 and recruits PLK4 to activate centriole biogenesis.
Specifications
| CCDC67 | |
| Polyclonal | |
| Unconjugated | |
| Deup1 | |
| 4933401K09Rik; CCDC67; coiled-coil domain containing 67; coiled-coil domain-containing protein 67; Deup1; deuterosomal protein 1; deuterosome assembly protein 1; deuterosome protein 1; RGD1306632 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 159989 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q05D60 | |
| Deup1 | |
| Recombinant protein corresponding to Human CCDC67. Recombinant protein control fragment (Product #RP-90402). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction