missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC66 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£181.00 - £255.00
Specifications
| Antigen | CCDC66 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663606
|
Novus Biologicals
NBP3-05878-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676385
|
Novus Biologicals
NBP3-05878-50ul |
50 μg |
£181.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCDC66 Polyclonal antibody specifically detects CCDC66 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CCDC66 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| coiled-coil domain containing 66, coiled-coil domain-containing protein 66, DKFZp686C0433 | |
| Produced in rabbits immunized with E. coli-derived Human CCDC66 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 285331 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title