missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581152
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CCDC6.
This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.
Specifications
| CCDC6 | |
| Polyclonal | |
| Unconjugated | |
| CCDC6 | |
| 2810012H18Rik; AA536681; AA960498; AW061011; CCDC6; coiled-coil domain containing 6; coiled-coil domain-containing protein 6; D10S170; H4; Papillary thyroid carcinoma-encoded protein; Protein H4; PTC; TPC; TST1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8030 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q16204 | |
| CCDC6 | |
| A synthetic peptide corresponding to the C-terminus of the Human CCDC6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction