missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC53 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100429
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CCDC53.
Acts as a component of the WASH core complex that functions as a nucleation-promoting factor (NPF) at the surface of endosomes, where it recruits and activates the Arp2/3 complex to induce actin polymerization, playing a key role in the fission of tubules that serve as transport intermediates during endosome sorting. [UniProt].
Specifications
| CCDC53 | |
| Polyclonal | |
| Unconjugated | |
| WASHC3 | |
| 2900091E11Rik; 5730495F03Rik; AD-016; Ccdc53; CGI-116; coiled-coil domain containing 53; coiled-coil domain-containing protein 53; RGD1563761; WASH complex subunit 3; WASH complex subunit CCDC53; WASHC3; x0009 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 299707, 51019, 67282 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CR27, Q9Y3C0 | |
| WASHC3 | |
| A synthesized peptide derived from human WASHC3(Accession Q9Y3C0), corresponding to amino acid residues V143-S193. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction