missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC51 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552831
This item is not returnable.
View return policy
Description
Immunogen sequence: RLQELKALLL EAQKGPVSLQ EAIREQASSY SRQQRDLHNL MVDLRGLVHA AGPGQDSGSQ AGSPPTRDRD VDVLSAALKE QLSHSRQVHS CLEGLREQLD GLEKTCSQMA GVVQLVKSAA HPGLVEPADG AMPSFLLEQG SMILAL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
Specifications
| CCDC51 | |
| Polyclonal | |
| Unconjugated | |
| CCDC51 | |
| 5730568A12Rik; AI551049; CCDC51; coiled-coil domain containing 51; coiled-coil domain-containing protein 51; RGD1311466 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96ER9 | |
| CCDC51 | |
| Recombinant protein corresponding to Human CCDC51. Recombinant protein control fragment (Product #RP-90390). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction