missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92355-0.02ml
This item is not returnable.
View return policy
Description
CCDC5 Polyclonal antibody specifically detects CCDC5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CCDC5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CCDC5Enhancer of invasion-cluster, coiled-coil domain containing 5 (spindle associated), FLJ40084, HAUS augmin-like complex subunit 1, HAUS augmin-like complex, subunit 1, HEI-CCoiled-coil domain-containing protein 5, HEICFLJ21094, HsT1461 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-250 of human HAUS1 (NP_612452.1). NDLTSDLFRTKSKSEEIKIELEKLEKNLTATLVLEKCLQEDVKKAELHLSTERAKVDNRRQNMDFLKAKSEEFRFGIKAAEEQLSARGMDASLSHQSLVALSEKLARLKQQTIPLKKKLESYLDLMPNPSL | |
| 0.02 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 115106 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction