missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCDC47 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12808982
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808982 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808982 Supplier Invitrogen™ Supplier No. PA520856

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse heart tissue lysate. PA5-20856 can be used with blocking peptide PEP-0970.

The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. The CCDC47 gene maps to chromosome 17 at 17q23.3. Little is known about this single-pass membrane protein except that the coiled-coil domain is within the cytosolic domain near the carboxy terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC47
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ccdc47
Gene Accession No. Q96A33, Q9D024
Gene Alias 2610204L23Rik; Adipocyte-specific protein 4; asp4; C88307; calumin; ccd47; CCDC47; coiled-coil domain containing 47; coiled-coil domain-containing protein 47; GK001; MSTP041; PSEC0077; RGD1308813
Gene Symbols Ccdc47
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human CCDC47.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57003, 67163
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.