missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCDC47 Polyclonal antibody specifically detects CCDC47 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CCDC47 |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
| Gene Alias | coiled-coil domain containing 47, coiled-coil domain-containing protein 47, GK001, MSTP041, PSEC0077 |
| Immunogen | CCDC47 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human CCDC47. The immunogen is located within amino acids 350 - 400 of CCDC47. Amino Acid Squence: NVPGSGNTYPKDMEAL |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 57003 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?