missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC47 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82077DL594
This item is not returnable.
View return policy
Description
CCDC47 Polyclonal antibody specifically detects CCDC47 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CCDC47 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| coiled-coil domain containing 47, coiled-coil domain-containing protein 47, GK001, MSTP041, PSEC0077 | |
| CCDC47 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CCDC47. The immunogen is located within the last 50 amino acids of CCDC47. Amino Acid Squence: RREQKKLEKKQMKMK | |
| 0.1 mL | |
| Cell Biology | |
| 57003 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction