missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC46 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | CCDC46 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CCDC46 Polyclonal antibody specifically detects CCDC46 in Human, Rat samples. It is validated for Western BlotSpecifications
| CCDC46 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 201134 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| coiled-coil domain containing 46, coiled-coil domain-containing protein 46, FLJ39610, MGC33887 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human CEP112 (NP_659473.2). MEVGSEEEKWEKLDAEFDHFVVDMKPFVLKLPHRTERQRCALWIRKLCEPSGTGAGIMGRKNRNLYAKLLLHMLKRGALEGPFTHRPEPGTLKILPSYMSIYFDEPNPARAKGSSPEGLPAWVLGELETSEHKLNESWKLSSGEDNTLVQSPTDVYSREQYTGKLRVRSHSLSPTHREDGQNITPKICEVYSKKSPVSLDDSDIEARLNSWNLGIENPRYLRQKPIPVSLMTPKFSLRKS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title