missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112096
This item is not returnable.
View return policy
Description
This antibody reacts with: Human, Mouse (Species predicted to react based on 100% sequence homology).
CCDC42 is required for sperm development.
Specifications
| CCDC42 | |
| Polyclonal | |
| PBS with no preservative | |
| Q96M95 | |
| CCDC42 | |
| A synthetic peptide corresponding to the center region of the Human CCDC42. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CCDC42 | |
| A530001H01Rik; CCDC42; CCDC42A; coiled-coil domain containing 42; coiled-coil domain-containing protein 42; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42; RGD1565925 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 146849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction