missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC42 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06102PCP
This item is not returnable.
View return policy
Description
CCDC42 Polyclonal antibody specifically detects CCDC42 in Human samples. It is validated for Western Blot
Specifications
| CCDC42 | |
| Polyclonal | |
| Western Blot | |
| CCDC42A, coiled-coil domain containing 42, coiled-coil domain-containing protein 42A, FLJ32734 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CCDC42. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 146849 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction