missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545994
This item is not returnable.
View return policy
Description
Peptide sequence: QELSITQSLC KARERETESE EHFKAIAQRE LGRVKDEIQR LENEMASILE Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.
The function of this protein remains unknown.
Specifications
| CCDC39 | |
| Polyclonal | |
| Unconjugated | |
| Ccdc39 | |
| 4921507O14Rik; AI447426; AI844750; b2b1304Clo; b2b1735Clo; b2b2025.1Clo; CCDC39; CILD14; coiled-coil domain containing 39; coiled-coil domain-containing protein 39; D3Ertd789e; FAP59; I79_014225; RGD1306277 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 339829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UFE4 | |
| Ccdc39 | |
| Synthetic peptide directed towards the N-terminal of human CCDC39 (aa 64-113). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction