missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561072
This item is not returnable.
View return policy
Description
Immunogen sequence: GEFIEMKSNL KHLEVLVAQQ SQEFQQLCEQ LGQLNVPSVL AELKRLISVP PVKDSASQTS PPLAQSLNLT RQEKYTSEKP VLWQAQALPA AWNPGMGSLQ Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 61%.
Required for DNA double-strand breaks (DSBs) formation in unsynapsed regions during meiotic recombination. Probably acts by forming a complex with MEI4 and REC114, which activates DSBs formation in unsynapsed regions, an essential step to ensure completion of synapsis. Not required for HORMAD1 functions in pairing-independent synaptonemal complex formation, ATR recruitment to unsynapsed axes, meiotic silencing of unsynapsed chromatin (MSUC) or meiotic surveillance.
Specifications
| CCDC36 | |
| Polyclonal | |
| Unconjugated | |
| CCDC36 | |
| Cancer/testis antigen 74; Ccdc36; coiled-coil domain containing 36; coiled-coil domain-containing protein 36; CT74; IHO1; Interactor of HORMAD1 protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IYA8 | |
| CCDC36 | |
| Recombinant protein corresponding to Human CCDC36. Recombinant protein control fragment (Product #RP-100228). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction