missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC36 Antibody (OTI2D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72504CL1
This item is not returnable.
View return policy
Description
CCDC36 Monoclonal antibody specifically detects CCDC36 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| CCDC36 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cancer/testis antigen 74, coiled-coil domain containing 36, coiled-coil domain-containing protein 36, CT74FLJ25320 | |
| Full length human recombinant protein of human CCDC36(NP_835467) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI2D11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 339834 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction