missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CCDC28A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29355
This item is not returnable.
View return policy
Description
CCDC28A Polyclonal antibody specifically detects CCDC28A in Human samples. It is validated for Immunoprecipitation
Specifications
| CCDC28A | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25901 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| C6orf80, CCRL1APMGC131913, chemokine C-C motif receptor-like 1 adjacent, chromosome 6 open reading frame 80, coiled-coil domain containing 28A, coiled-coil domain-containing protein 28A, DKFZp586D0623 | |
| between 100 and 150 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction