missing translation for 'onlineSavingsMsg'
Learn More

CCDC21 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18416007 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416007 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416007 Supplier Novus Biologicals Supplier No. NBP178219R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC21 Polyclonal antibody specifically detects CCDC21 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC21
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias coiled-coil domain containing 21, coiled-coil domain-containing protein 21, DKFZp434L0117, DKFZp434P232, FLJ13976, FLJ22000
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 675 and 725 of human Centrosomal Protein 85kDa using the numbering given in entry NP_073615.2 (GeneID 64793).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 64793
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.