missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC21 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | CCDC21 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CCDC21 Polyclonal antibody specifically detects CCDC21 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CCDC21 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 64793 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| coiled-coil domain containing 21, coiled-coil domain-containing protein 21, DKFZp434L0117, DKFZp434P232, FLJ13976, FLJ22000 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-430 of human CEP85 (NP_073615.2). TRNGDLGAMKHSPGLSRDLMYFSGATGENGIEQSWFPAVGHERQEEARKFDIPSMESTLNQSAMMETLYSDPHHRVRFHNPRTSTSKELYRVLPEAKKAPGSGAVFERNGPHSNSSGVLPLGLQPAPGLSKPLPSQVWQPSPDTWHPREQSCELSTCRQQLELIRLQMEQMQLQNGAICHHPAAFGPSLPILEPAQWISILNSNEHLLKEKELLIDKQRKHISQLEQKVRESELQVHSALLGRPAPFGDVCLLRLQELQRENTFLRAQFAQKTEALSREKIDLEKKLSASEVEVQLIRESL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title