missing translation for 'onlineSavingsMsg'
Learn More

CCDC190 Antibody, Novus Biologicals™

Product Code. 18415131 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415131 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415131 Supplier Novus Biologicals Supplier No. NBP182674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC190 Polyclonal antibody specifically detects CCDC190 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC190
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:20 to 1:50, Immunohistochemistry-Paraffin 1:20 to 1:50
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias C1orf110, chromosome 1 open reading frame 110, Coiled-Coil Domain Containing 190, FLJ41579, MGC48998, RP11-331H2.2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: KDVDPSKGISVPCQNQEVSTNTIEQGPSSSPASDSGMACADETRSKDVALKPDGNTGKQI
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339512
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.