missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550886
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CFAP45 protein.
The function remains unknown.
Specifications
| CCDC19 | |
| Polyclonal | |
| Unconjugated | |
| CFAP45 | |
| 1700028D05Rik; CCDC19; CFAP45; cilia and flagella associated protein 45; cilia- and flagella-associated protein 45; coiled-coil domain containing 19; Coiled-coil domain-containing protein 19; coiled-coil domain-containing protein 19, mitochondrial; nasopharyngeal epithelium specific protein 1; nasopharyngeal epithelium specific protein 1 (NESG1); nasopharyngeal epithelium-specific protein 1; NESG1; protein CFAP45, mitochondrial; Unknown (protein for MGC:134208) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25790 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UL16 | |
| CFAP45 | |
| Synthetic peptide corresponding to residues near the N terminal of human cilia and flagella associated protein 45. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction