missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC18 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30024-1-AP-150UL
This item is not returnable.
View return policy
Description
The CCDC18 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CCDC18. This antibody recognizes human, rat antigen. The CCDC18 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| CCDC18 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 343099 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 500 μg/mL | |
| NY-SAR-41; RP4-717I23.1; dJ717I23.1 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| 151 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction