missing translation for 'onlineSavingsMsg'
Learn More

CCDC18 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18420695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420695 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420695 Supplier Novus Biologicals Supplier No. NBP178738UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC18 Polyclonal antibody specifically detects CCDC18 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC18
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias coiled-coil domain containing 18, coiled-coil domain-containing protein 18, dJ717I23.1, NY-SAR-41, sarcoma antigen NY-SAR-41
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 225 and 275 of human Coiled-Coil Domain Containing 18 using the numbering given in entry CAI22508.1 (GeneID 343099).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343099
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.