missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC176 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551833
This item is not returnable.
View return policy
Description
Immunogen sequence: ISRKHYKQIA QAAFNLKMRA ACTGRTEYPK IRTFDGREHS TNSVNQDLLE AEKWTHIEGN VDIGDLTWEQ KEKVLRLLFA KMNGCPSRKY NQSSRPPVPD YVVSDSGETK EFGDESKLQD KIFITQQIAI SDSSGEVVLP TIPKEPQ Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
Basal body protein required in multiciliate cells to align and maintain cilia orientation in response to flow. May act by mediating a maturation step that stabilizes and aligns cilia orientation. Not required to respond to planar cell polarity (PCP) or flow-based orientation cues.
Specifications
| CCDC176 | |
| Polyclonal | |
| Unconjugated | |
| BBOF1 | |
| 2900006K08Rik; AI427784; basal body orientation factor 1; basal body-orientation factor 1; BBOF1; C10H14orf45; C14orf45; C8H14orf45; Ccdc176; coiled-coil domain containing 176; coiled-coil domain-containing protein 176; RGD1560978; uncharacterized protein C14orf45 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8ND07 | |
| BBOF1 | |
| Recombinant protein corresponding to Human CCDC176. Recombinant protein control fragment (Product #RP-89083). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction