missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC159 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570972
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
CCDC159 is a protein coding gene.
Specifications
| CCDC159 | |
| Polyclonal | |
| Unconjugated | |
| CCDC159 | |
| CCDC159; coiled-coil domain containing 159; coiled-coil domain-containing protein 159 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 126075 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P0C7I6 | |
| CCDC159 | |
| synthetic peptide directed towards the C-terminal region of human CCDC159. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction