missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC149B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572621
This item is not returnable.
View return policy
Description
CCDC149B Polyclonal Antibody for Western Blot
Specifications
| CCDC149B | |
| Polyclonal | |
| Unconjugated | |
| ccdc149b | |
| ccdc149; ccdc149a; ccdc149b; coiled-coil domain containing 149; coiled-coil domain containing 149b; coiled-coil domain-containing protein 149-A; coiled-coil domain-containing protein 149-B; fi16d12; wu:fi16d12; zgc:92601 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 436983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6DH86 | |
| ccdc149b | |
| KLH conjugated synthetic peptide between 62-89 amino acids from the N-terminal region of DANRE ccdc149b. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction