missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC144B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | CCDC144B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CCDC144B Polyclonal specifically detects CCDC144B in Human samples. It is validated for Western Blot.Specifications
| CCDC144B | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 284047 | |
| Synthetic peptide directed towards the middle region of human CCDC144B. Peptide sequence TELSGTLTDGTTVGNDDDGLNQQIPRKENEEHDRPADKTANEKNKVKNQI. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CCDC144B coiled-coil domain containing 144B (pseudogene) | |
| CCDC144B | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title