missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC137 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92389-0.1ml
This item is not returnable.
View return policy
Description
CCDC137 Polyclonal antibody specifically detects CCDC137 in Human samples. It is validated for Western Blot
Specifications
| CCDC137 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| coiled-coil domain containing 137, coiled-coil domain-containing protein 137, MGC16597 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 213-288 of human CCDC137 (NP_954981.1). QRSVSKDQPGRRSQMLRMLLSPGGVSQPLTASLARQRIVEEERERAVQAYRALKQRQQQLHGERPHLTSRKKPEPQ | |
| 0.1 mL | |
| Cell Biology | |
| 339230 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction