missing translation for 'onlineSavingsMsg'
Learn More

CCDC134 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18334736 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334736 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334736 Supplier Novus Biologicals Supplier No. NBP300083AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC134 Polyclonal specifically detects CCDC134 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC134
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias coiled-coil domain containing 134, coiled-coil domain-containing protein 134, dJ821D11.3, FLJ22349, MGC21013
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CCDC134 (Q9H6E4; Met1-Leu229).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79879
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.