missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC134 Antibody (026), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90189AF647
This item is not returnable.
View return policy
Description
CCDC134 Monoclonal specifically detects CCDC134 in Human samples. It is validated for Western Blot.
Specifications
| CCDC134 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79879 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 26 | |
| Western Blot | |
| coiled-coil domain containing 134, coiled-coil domain-containing protein 134, dJ821D11.3, FLJ22349, MGC21013 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCDC134 (Q9H6E4; Met1-Leu229). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction