missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC124 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92737-0.1ml
This item is not returnable.
View return policy
Description
CCDC124 Polyclonal antibody specifically detects CCDC124 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CCDC124 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| coiled-coil domain containing 124, coiled-coil domain-containing protein 124 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-223 of human CCDC124 (NP_612451.1). MPKKFQGENTKSAAARARRAEAKAAADAKKQKELEDAYWKDDDKHVMRKEQRKEEKEKRRLDQLERKKETQRLLEEEDSKLKGGKAPRVATSSKVTRAQIEDTLRRDHQLREAPDTAEKAKSHLEVPLEENVNRRVLEEGSVEARTIEDAIAVLSVAEEAADRHPERRMRAAFTAFEEAQLPRLKQENPNMRLSQLKQLLKKEWLRSPDNPMNQRAVPFNAPK | |
| 0.1 mL | |
| Cell Biology | |
| 115098 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction