missing translation for 'onlineSavingsMsg'
Learn More

CCDC110 Antibody, Novus Biologicals™

Product Code. p-7109431 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18210826 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18210826 Supplier Novus Biologicals Supplier No. NBP179593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC110 Polyclonal specifically detects CCDC110 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC110
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_689988
Gene Alias Cancer/testis antigen 52, coiled-coil domain containing 110, coiled-coil domain-containing protein 110, CT52Cancer/testis antigen KM-HN-1, KMHN1KM-HN-1, MGC33607
Gene Symbols CCDC110
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CCDC110The immunogen for this antibody is CCDC110. Peptide sequence KEELKKHSQENIKFENSISRLTEDKILLENYVRSIENERDTLEFEMRHLQ.
Molecular Weight of Antigen 97 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256309
Test Specificity Expected identity based on immunogen sequence: Canine: 86%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.