Learn More
Invitrogen™ CCDC11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595816
Description
Immunogen sequence: MYSQRFGTVQ REVKGPTPKV VIVRSKPPKG QGAEHHLERI RRSHQKHNAI LASIKSSERD RLKAEWDQHN DCKILDSLVR ARIKDAVQGF IINIEERRNK LRELLALEEN EYFTEMQLKK ETIEEKKDRM REKTKLLKEK NEKERQDFVA EKLDQQFRER CEELRVELLS IHQKKVCEER KAQIAFNEEL SRQKLVEEQM FSKLWEEDRL AKEKREAQEA RRQKELMENT RLGLNAQITS IKAQRQATQL LKEEEARLVE SNNAQIKHEN EQDMLKKQKA KQETRTILQK ALQERIEHIQ; Positive Samples: Mouse testis, Rat testis; Cellular Location: Cell projection, cilium.
This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning. [provided by RefSeq, Aug 2015].
Specifications
| CCDC11 | |
| Polyclonal | |
| Unconjugated | |
| CFAP53 | |
| 4933415I03Rik; Ccdc11; Cfap53; cilia and flagella associated protein 53; cilia- and flagella-associated protein 53; coiled-coil domain containing 11; coiled-coil domain-containing protein 11; HTX6; mitogen-activated protein kinase kinase kinase kinase 4-like; RGD1306734 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 220136, 364899, 74453 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.85 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96M91, Q9D439 | |
| CFAP53 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CFAP53 (NP_6594572). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.