missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC106 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-21003 can be used with blocking peptide PEP-1117.
The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. CCDC106 was initially identified as a p53-interacting protein by yeast two-hybrid screening. Other experiments demonstrated that CCDC106 co-localizes and interacts with p53 in the nucleus, inhibiting the transcriptional activity of p53 and stimulating p53 protein degradation, indicating that at least one of the functions of CCDC106 is acting as a negative regulator of p53.
Specifications
Specifications
| Antigen | CCDC106 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CCDC106 |
| Gene Accession No. | Q3ULM0, Q9BWC9 |
| Gene Alias | BC018462; CCDC106; coiled-coil domain containing 106; coiled-coil domain-containing protein 106; HSU79303; MNCb-2071; protein predicted by clone 23882; RGD1566222; ZNF581 |
| Gene Symbols | CCDC106 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?