missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC106 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77381C
This item is not returnable.
View return policy
Description
CCDC106 Polyclonal antibody specifically detects CCDC106 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CCDC106 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| coiled-coil domain containing 106, coiled-coil domain-containing protein 106, HSU79303, protein predicted by clone 23882, ZNF581 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human CCDC106. The immunogen is located within the last 50 amino acids of CCDC106. Amino Acid Squence: CEFDPSKERLLEYSRR | |
| 0.1 mL | |
| Signal Transduction | |
| 29903 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction