missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC101 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£130.00 - £318.00
Specifications
| Antigen | CCDC101 |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:100-1:200 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18327855
|
Novus Biologicals
NBP3-18637-25UL |
25 μg |
£130.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18351664
|
Novus Biologicals
NBP3-18637-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCDC101 Polyclonal antibody specifically detects CCDC101 in Human samples. It is validated for Western Blot, ELISASpecifications
| CCDC101 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| Whole antiserum | |
| coiled-coil domain containing 101, Coiled-Coil Domain-Containing Protein 101, FLJ32446, SAGA-Associated Factor 29 Homolog, SGF29, STAF36 | |
| Polyclonal antibody raised in rabbit against human CCDC101 (coiled-coil domain containing 101), using two KLH-conjugated synthetic peptides containing a sequence from the central and from the C-terminal part of the protein, respectively. | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:100-1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q96ES7 | |
| 112869 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title