missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCDC101 Polyclonal antibody specifically detects CCDC101 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | CCDC101 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:200 |
| Formulation | Whole antiserum |
| Gene Accession No. | Q96ES7 |
| Gene Alias | coiled-coil domain containing 101, Coiled-Coil Domain-Containing Protein 101, FLJ32446, SAGA-Associated Factor 29 Homolog, SGF29, STAF36 |
| Host Species | Rabbit |
| Immunogen | Polyclonal antibody raised in rabbit against human CCDC101 (coiled-coil domain containing 101), using two KLH-conjugated synthetic peptides containing a sequence from the central and from the C-terminal part of the protein, respectively. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?