missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC93 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562935
This item is not returnable.
View return policy
Description
Immunogen sequence: QGMDFIHIFP VVQWLVKRAI ETKEEMGDYI RSYSVSQFQK TYSLPEDDDF IKRKEKAIKT VVDLSEVYKP RRKYKRHQGA EELLDEESRI HATLLEYG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.
CCDC93 is involved in copper-dependent ATP7A trafficking between the trans-Golgi network and vesicles in the cell periphery; the function is proposed to depend on its association within the CCC complex and cooperation with the WASH complex on early endosomes and is dependent on its interaction with WASHC2C.
Specifications
| CCDC93 | |
| Polyclonal | |
| Unconjugated | |
| CCDC93 | |
| 4633402D15Rik; 9230102M16Rik; CCDC93; coiled-coil domain containing 93; coiled-coil domain-containing protein 93 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q567U6 | |
| CCDC93 | |
| Recombinant protein corresponding to Human CCD93. Recombinant protein control fragment (Product #RP-99631). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction