missing translation for 'onlineSavingsMsg'
Learn More

CCCP, MedChemExpress

Product Code. 16459766
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459766 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459766 Supplier Medchem Express Supplier No. HY10094150mg
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CCCP is an oxidative phosphorylation (OXPHOS) uncoupler. CCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation.

CCCP is an oxidative phosphorylation (OXPHOS) uncoupler. CCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation.

Specifications

Chemical Name or Material CCCP
Physical Form Solid
CAS 555-60-2
Color Coffee
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C9H5ClN4
For Use With (Application) Cancer-Kinase/protease
Synonym Carbonyl cyanide 3-chlorophenylhydrazone Carbonyl Cyanide m-Chlorophenylhydrazone
Solubility Information DMSO : 50 mg/mL (244.36 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES N#C/C(C#N)=N/NC1=CC=CC(Cl)=C1
Molecular Weight (g/mol) 204.62
Formula Weight 204.62
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.