missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCBL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554342
This item is not returnable.
View return policy
Description
Immunogen sequence: LVDEGDEVII IEPFFDCYEP MTMMAGGRPV FVSLKPGPIQ NGELGSSSNW QLDPMELAGK FTSRTKALVL NTPNNPLGKV F Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
Kynurenine aminotransferases KAT I, KAT II, and KAT III belong to the class-I pyridoxal-phosphate-dependent aminotransferase family. KAT I is a cytoplasmic protein involved in glutamine catabolism. KAT I functions in the catalysis of the transamination of L-kinurenine to form kynurenic acid, a neuroprotective and anticonvulsant metabolite of tryptophan. Kynurenic acid is involved in synaptic transmission and has been implicated in a number of neurological disorders including schizophrenia and Huntington's disease. KAT I also functions in the metabolism of cysteine conjugates in some halogenated alkenes and alkanes to form reactive metabolites. KAT I has three isoforms. Isoform 1is the full length protein, isoform 2 lacks amino acids 68-117 and isoform 3 lacks amino acids 251-422. Based on sequence similarity, KAT I is thought to function as a homodimer.
Specifications
| CCBL1 | |
| Polyclonal | |
| Unconjugated | |
| KYAT1 | |
| 2010009K05Rik; AI182306; beta-lysase, kidney; CCBL1; cysteine conjugate beta lyase 1; cysteine conjugate-beta lyase 1; cysteine conjugate-beta lyase, cytoplasmic; cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); cysteine-S-conjugate beta-lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); glutamine transaminase K; glutamine-phenylpyruvate aminotransferase; Glutamine--phenylpyruvate transaminase; Gtk; Kat; KAT1; KATI; KYAT1; kyneurenine aminotransferase; kynurenine aminotransferase 1; Kynurenine aminotransferase I; kynurenine aminotransferase/glutamine transaminase K; kynurenine--oxoglutarate transaminase 1; Kynurenine--oxoglutarate transaminase 1, mitochondrial; kynurenine--oxoglutarate transaminase I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16773 | |
| KYAT1 | |
| Recombinant protein corresponding to Human CCBL1. Recombinant protein control fragment (Product #RP-93064). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction