missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CC2D2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100428
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CC2D2A.
This gene encodes a coiled-coil and calcium binding domain protein that appears to play a critical role in cilia formation. Mutations in this gene cause Meckel syndrome type 6, as well as Joubert syndrome type 9. Alternative splicing results in multiple transcript variants.
Specifications
| CC2D2A | |
| Polyclonal | |
| Unconjugated | |
| CC2D2A | |
| 5730509K17Rik; b2b1035Clo; CC2D2A; coiled-coil and C2 domain containing 2A; coiled-coil and C2 domain-containing protein 2A; JBTS9; KIAA1345; MKS6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 231214, 57545 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CFW7, Q9P2K1 | |
| CC2D2A | |
| A synthesized peptide derived from human CC2D2A(Accession Q9P2K1), corresponding to amino acid residues V997-I1047. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction