missing translation for 'onlineSavingsMsg'
Learn More

CC2D2A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-47705

Product Code. 30559294

  • £228.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CC2D2A Polyclonal antibody specifically detects CC2D2A in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CC2D2A
Polyclonal
Western Blot 1:500-1:1000, ELISA, Immunohistochemistry 1:20-1:200, Immunocytochemistry/ Immunofluorescence 1:20-1:200
coiled-coil and C2 domain containing 2A
coiled-coil and C2 domain containing 2A. (Uniprot# Q9P2K1)
100 μg
Primary
Human, Mouse
Purified
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS, 50% Glycerol, pH (7.3)
Rabbit
Immunogen affinity purified
RUO
57545
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.