missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CC2D1A Recombinant Rabbit Monoclonal Antibody (23GB3045), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CC2D1A |
|---|---|
| Clone | 23GB3045 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089007
![]() |
Invitrogen™
MA553304 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089304
|
Invitrogen™
MA553305 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CC2D1A Recombinant Monoclonal antibody specifically detects CC2D1A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CC2D1A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q66HA5, Q6P1N0, Q8K1A6 | |
| 212139, 288908, 54862 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3045 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5-HT1A receptor; AKI1; Akt kinase-interacting protein 1; BC016188; CC2D1A; coiled-coil and C2 domain containing 1A; coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five' repressor element under dual repression binding protein 1; five repressor element under dual repression-binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; Freud-1; Freud-1/Aki1; mental retardation, nonsyndromic, autosomal recessive, 3; MRT3; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein; RGD1306108; Tape | |
| A synthesized peptide derived from CC2D1A (750-951AA). | |
| Primary | |
| -20°C | |
| CC2D1A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title