missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC2D1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CC2D1A |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16876564
|
Proteintech
16816-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16866564
|
Proteintech
16816-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5′-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.Specifications
| CC2D1A | |
| Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q66HA5, Q6P1N0, Q8K1A6 | |
| 212139, 288908, 54862 | |
| CC2D1A Fusion Protein Ag10355 | |
| Primary | |
| -20°C | |
| CC2D1A |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CC2D1A, FREUD 1, Freud 1/Aki1, MRT3 | |
| Cc2d1a | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title